missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nonanophenone, 97%

Manufacturer:  Alfa Aesar™ A1223809

 View more versions of this product

Catalog No. AAA1223809



Specifications

Specifications

Nonanophenone
139°C to 140°C (2mmHg)
10g
97%
MFCD00048965
n-Octyl phenyl ketone
CCCCCCCCC(=O)C1=CC=CC=C1
218.34
218.34
>110°C (230°F)
14°C
6008-36-2
C15H22O
2047454
PFUPABFCHVRLLY-UHFFFAOYSA-N
1-phenylnonan-1-one
80108

Chemical Identifiers

6008-36-2
218.34
PFUPABFCHVRLLY-UHFFFAOYSA-N
80108
CCCCCCCCC(=O)C1=CC=CC=C1
C15H22O
MFCD00048965
n-Octyl phenyl ketone
1-phenylnonan-1-one
SDS Documents