Please login to your online account to display your discounted pricing

Alfa Aesar™ Nonanophenone, 97%

$105.78 - $394.77

Chemical Identifiers

Chemical Name or Material Nonanophenone
CAS 6008-36-2
Molecular Formula C15H22O
Formula Weight 218.34
MDL Number MFCD00048965
Beilstein 2047454
Synonym n-Octyl phenyl ketone
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1223809

  SDS
alfa aesar™
A1223809
10g Each for $105.78

AAA1223818

  SDS
alfa aesar™
A1223818
50g Each for $394.77
Description & Specifications

Specifications

Formula Weight 218.34
Melting Point 14°C
Assay 97%
Molecular Formula C15H22O
Synonym n-Octyl phenyl ketone
Quantity 10g
Chemical Name or Material Nonanophenone
CAS 6008-36-2
Beilstein 2047454
Flash Point >110°C (230°F)
MDL Number MFCD00048965
Boiling Point 139° to 140°C (2mmHg)