missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nonanoic Acid, TCI America™

$21.39 - $56.53

Chemical Identifiers

CAS 112-05-0
Molecular Formula C9H18O2
Molecular Weight (g/mol) 158.241
MDL Number MFCD00004433
InChI Key FBUKVWPVBMHYJY-UHFFFAOYSA-N
Synonym pelargonic acid, n-nonanoic acid, nonylic acid, pelargic acid, nonoic acid, n-nonylic acid, n-nonoic acid, 1-octanecarboxylic acid, pelargon, cirrasol 185a
PubChem CID 8158
ChEBI CHEBI:29019
IUPAC Name nonanoic acid
SMILES CCCCCCCCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N0288500ML View Documents TCI America
N0288500ML
500mL Each for $56.53
 
N028825ML View Documents TCI America
N028825ML
25mL Each for $21.39
 
Specifications

Chemical Identifiers

112-05-0
158.241
FBUKVWPVBMHYJY-UHFFFAOYSA-N
8158
nonanoic acid
C9H18O2
MFCD00004433
pelargonic acid, n-nonanoic acid, nonylic acid, pelargic acid, nonoic acid, n-nonylic acid, n-nonoic acid, 1-octanecarboxylic acid, pelargon, cirrasol 185a
CHEBI:29019
CCCCCCCCC(=O)O

Specifications

254°C
500mL
C9H18O2
3265
FBUKVWPVBMHYJY-UHFFFAOYSA-N
nonanoic acid
8158
158.24
≥90.0% (GC)
Yellow
112-05-0
MFCD00004433
pelargonic acid, n-nonanoic acid, nonylic acid, pelargic acid, nonoic acid, n-nonylic acid, n-nonoic acid, 1-octanecarboxylic acid, pelargon, cirrasol 185a
CCCCCCCCC(=O)O
158.241
CHEBI:29019
Clear Liquid at 20°C
Nonanoic Acid
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0608

RTECSNumber : RA6650000

TSCA : Yes