missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nonanal Diethyl Acetal, TCI America™

$59.58 - $186.39

Chemical Identifiers

CAS 54815-13-3
Molecular Formula C13H28O2
Molecular Weight (g/mol) 216.365
MDL Number MFCD00672806
InChI Key RJMSGTVQHHFVLK-UHFFFAOYSA-N
Synonym 1,1-Diethoxynonane, Nonyl Aldehyde Diethyl Acetal
PubChem CID 108625
IUPAC Name 1,1-diethoxynonane
SMILES CCCCCCCCC(OCC)OCC
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N09565ML View Documents TCI America
N09565ML
5mL Each for $59.58
 
N095625ML View Documents TCI America
N095625ML
25mL Each for $186.39
 
Specifications

Chemical Identifiers

54815-13-3
216.365
RJMSGTVQHHFVLK-UHFFFAOYSA-N
108625
CCCCCCCCC(OCC)OCC
C13H28O2
MFCD00672806
1,1-Diethoxynonane, Nonyl Aldehyde Diethyl Acetal
1,1-diethoxynonane

Specifications

66°C
5mL
C13H28O2
1,1-Diethoxynonane, Nonyl Aldehyde Diethyl Acetal
CCCCCCCCC(OCC)OCC
216.365
216.37
≥98.0% (GC)
Colorless
54815-13-3
MFCD00672806
RJMSGTVQHHFVLK-UHFFFAOYSA-N
1,1-diethoxynonane
108625
Clear Liquid at 20°C
Nonanal Diethyl Acetal
Safety and Handling

Safety and Handling

TSCA : Yes