missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nonanal Diethyl Acetal 98.0+%, TCI America™

Manufacturer:  TCI America N095625ML

 View more versions of this product

Catalog No. N095625ML



Specifications

Specifications

Nonanal Diethyl Acetal
Colorless
54815-13-3
MFCD00672806
RJMSGTVQHHFVLK-UHFFFAOYSA-N
1,1-diethoxynonane
108625
Clear Liquid at 20°C
66°C
25mL
C13H28O2
1,1-Diethoxynonane, Nonyl Aldehyde Diethyl Acetal
CCCCCCCCC(OCC)OCC
216.365
216.37
≥98.0% (GC)

Chemical Identifiers

54815-13-3
216.365
RJMSGTVQHHFVLK-UHFFFAOYSA-N
108625
CCCCCCCCC(OCC)OCC
C13H28O2
MFCD00672806
1,1-Diethoxynonane, Nonyl Aldehyde Diethyl Acetal
1,1-diethoxynonane
Safety and Handling

Safety and Handling

TSCA : Yes

Documents