Please login to your online account to display your discounted pricing

Alfa Aesar™ Nonafluoro-2-iodobutane, 98%, stab. with copper

$63.73 - $255.53

Chemical Identifiers

Chemical Name or Material Nonafluoro-2-iodobutane
CAS 375-51-9
Name Note stabilized with copper
Molecular Formula CF3CF2CFICF3
Formula Weight 345.93
MDL Number MFCD00077556
Beilstein 1777547
Synonym 2-Iodononafluorobutane; Perfluoroisobutyl iodide
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAL1679206

  SDS
alfa aesar™
L1679206
5g Each for $63.73

AAL1679214

  SDS
alfa aesar™
L1679214
25g Each for $255.53
Description & Specifications

Specifications

Formula Weight 345.93
Chemical Name or Material Nonafluoro-2-iodobutane
Molecular Formula CF3CF2CFICF3
Synonym 2-Iodononafluorobutane; Perfluoroisobutyl iodide
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Boiling Point 64° to 67°C
Name Note stabilized with copper
Assay 98%
MDL Number MFCD00077556
CAS 375-51-9
Quantity 5g
Beilstein 1777547
Health Hazard 3 GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage 2° to 8°C
Safety and Handling

Warning

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

2° to 8°C