missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nonafluoro-1-butanesulfonic Acid, TCI America™

$80.97 - $282.64

Chemical Identifiers

CAS 375-73-5
Molecular Formula C4HF9O3S
Molecular Weight (g/mol) 300.095
MDL Number MFCD01320794
InChI Key JGTNAGYHADQMCM-UHFFFAOYSA-N
Synonym Perfluoro-1-butanesulfonic Acid
PubChem CID 67815
IUPAC Name 1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonic acid
SMILES C(C(C(F)(F)S(=O)(=O)O)(F)F)(C(F)(F)F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N070925G View Documents TCI America
N070925G
25g Each for $282.64
 
N07095G View Documents TCI America
N07095G
5g Each for $80.97
 
Specifications

Chemical Identifiers

375-73-5
300.095
JGTNAGYHADQMCM-UHFFFAOYSA-N
67815
C(C(C(F)(F)S(=O)(=O)O)(F)F)(C(F)(F)F)(F)F
C4HF9O3S
MFCD01320794
Perfluoro-1-butanesulfonic Acid
1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonic acid

Specifications

212°C
25g
C4HF9O3S
2586
JGTNAGYHADQMCM-UHFFFAOYSA-N
1,1,2,2,3,3,4,4,4-nonafluorobutane-1-sulfonic acid
67815
Clear Liquid at 20°C
Nonafluoro-1-butanesulfonic Acid
Yellow
375-73-5
MFCD01320794
Perfluoro-1-butanesulfonic Acid
C(C(C(F)(F)S(=O)(=O)O)(F)F)(C(F)(F)F)(F)F
300.095
300.09
≥98.0% (T)
Safety and Handling

Safety and Handling

RTECSNumber : EK5930000

TSCA : Yes