missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nonadecylcyclohexane, TCI America™

$279.59 - $931.97

Chemical Identifiers

CAS 22349-03-7
Molecular Formula C25H50
Molecular Weight (g/mol) 350.675
MDL Number MFCD00058954
InChI Key YVTPTBVYGTYTKP-UHFFFAOYSA-N
Synonym 1-Cyclohexylnonadecane
PubChem CID 89671
IUPAC Name nonadecylcyclohexane
SMILES CCCCCCCCCCCCCCCCCCCC1CCCCC1
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N039425G View Documents TCI America
N039425G
25g Each for $931.97
 
N03945G View Documents TCI America
N03945G
5g Each for $279.59
 
Specifications

Chemical Identifiers

22349-03-7
350.675
YVTPTBVYGTYTKP-UHFFFAOYSA-N
89671
CCCCCCCCCCCCCCCCCCCC1CCCCC1
C25H50
MFCD00058954
1-Cyclohexylnonadecane
nonadecylcyclohexane

Specifications

420°C
45°C
22349-03-7
MFCD00058954
YVTPTBVYGTYTKP-UHFFFAOYSA-N
nonadecylcyclohexane
89671
Crystal-Powder at 20°C
Nonadecylcyclohexane
White-Yellow
25g
C25H50
1-Cyclohexylnonadecane
CCCCCCCCCCCCCCCCCCCC1CCCCC1
350.675
350.68
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No