Please login to your online account to display your discounted pricing

Nonadecanoic acid, 98+%, ACROS Organics™


Quantity 1g
Packaging Glass bottle

Manufacturer: fisher scientific acros  AC207190010

 View more versions of this product

Catalog No. AC207190010

  • / Each
 

Description & Specifications

Chemical Identifiers

Assay ≥98.0%
Formula Weight 298.51
Physical Form Fine Powder or Shiny Flakes
Quantity 1g
Color White
Beilstein 02, II, 368
CAS Min % 98.0
CAS Max % 100.0
Packaging Glass bottle
Infrared Spectrum Authentic
Linear Formula CH3(CH2)17CO2H
Health Hazard 2 GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
Health Hazard 3 GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Chemical Name or Material Nonadecanoic acid
MDL Number MFCD00002754
CAS 646-30-0
Molecular Formula C19H38O2
Assay Percent Range 98+%

Specifications

Chemical Name or Material Nonadecanoic acid
CAS 646-30-0
CAS Min % 98.0
CAS Max % 100.0
Assay ≥98.0%
Assay Percent Range 98+%
Molecular Formula C19H38O2
Linear Formula CH3(CH2)17CO2H
Formula Weight 298.51
MDL Number MFCD00002754
Accessories

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.

SDS & Safety Documents