missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nonadecanedioic Acid, TCI America™

$452.23 - $1,492.67

Chemical Identifiers

CAS 6250-70-0
Molecular Formula C19H36O4
Molecular Weight (g/mol) 328.493
MDL Number MFCD00039536
InChI Key IFAWYXIHOVRGHQ-UHFFFAOYSA-N
Synonym 1,17-Heptadecanedicarboxylic Acid
PubChem CID 12572015
IUPAC Name nonadecanedioic acid
SMILES C(CCCCCCCCC(=O)O)CCCCCCCCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N06631G View Documents TCI America
N06631G
1g Each for $452.23
 
N06635G View Documents TCI America
N06635G
5g Each for $1,492.67
 
Specifications

Chemical Identifiers

6250-70-0
328.493
IFAWYXIHOVRGHQ-UHFFFAOYSA-N
12572015
C(CCCCCCCCC(=O)O)CCCCCCCCC(=O)O
C19H36O4
MFCD00039536
1,17-Heptadecanedicarboxylic Acid
nonadecanedioic acid

Specifications

Undesignated
6250-70-0
MFCD00039536
IFAWYXIHOVRGHQ-UHFFFAOYSA-N
nonadecanedioic acid
12572015
Crystal-Powder at 20°C
Nonadecanedioic Acid
1g
C19H36O4
1,17-Heptadecanedicarboxylic Acid
C(CCCCCCCCC(=O)O)CCCCCCCCC(=O)O
328.493
328.49
≥98.0% (T)
Safety and Handling

Safety and Handling

TSCA : No