missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nonadecafluorodecanenitrile, TCI America™

$152.78 - $453.77

Chemical Identifiers

CAS 379215-40-4
Molecular Formula C10F19N
Molecular Weight (g/mol) 495.087
MDL Number MFCD00045610
InChI Key QXBUHLMUCFBZOL-UHFFFAOYSA-N
Synonym Perfluorodecanonitrile, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Nonadecafluorodecanenitrile
PubChem CID 2776067
IUPAC Name 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluorodecanenitrile
SMILES C(#N)C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N10951G View Documents TCI America
N10951G
1g Each for $152.78
 
N10955G View Documents TCI America
N10955G
5g Each for $453.77
 
Specifications

Chemical Identifiers

379215-40-4
495.087
QXBUHLMUCFBZOL-UHFFFAOYSA-N
2776067
C(#N)C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
C10F19N
MFCD00045610
Perfluorodecanonitrile, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Nonadecafluorodecanenitrile
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluorodecanenitrile

Specifications

Yellow
379215-40-4
MFCD00045610
QXBUHLMUCFBZOL-UHFFFAOYSA-N
2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-nonadecafluorodecanenitrile
2776067
Clear Liquid at 20°C
Nonadecafluorodecanenitrile
1g
C10F19N
Perfluorodecanonitrile, 2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,10-Nonadecafluorodecanenitrile
C(#N)C(C(C(C(C(C(C(C(C(F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F)(F)F
495.087
495.09
≥98.0% (GC)
Safety and Handling

Safety and Handling

TSCA : No