missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nonacosanoic Acid, TCI America™

$418.63 - $1,462.11

Chemical Identifiers

CAS 4250-38-8
Molecular Formula C29H58O2
Molecular Weight (g/mol) 438.781
MDL Number MFCD00057830
InChI Key IHEJEKZAKSNRLY-UHFFFAOYSA-N
PubChem CID 20245
ChEBI CHEBI:84867
IUPAC Name nonacosanoic acid
SMILES CCCCCCCCCCCCCCCCCCCCCCCCCCCCC(=O)O
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
N06625G View Documents TCI America
N06625G
5g Each for $1,462.11
 
N06621G View Documents TCI America
N06621G
1g Each for $418.63
 
Specifications

Chemical Identifiers

4250-38-8
438.781
IHEJEKZAKSNRLY-UHFFFAOYSA-N
CHEBI:84867
CCCCCCCCCCCCCCCCCCCCCCCCCCCCC(=O)O
C29H58O2
MFCD00057830
20245
nonacosanoic acid

Specifications

White
5g
C29H58O2
IHEJEKZAKSNRLY-UHFFFAOYSA-N
nonacosanoic acid
20245
438.78
≥95.0% (GC)
90°C
4250-38-8
MFCD00057830
CCCCCCCCCCCCCCCCCCCCCCCCCCCCC(=O)O
438.781
CHEBI:84867
Crystal-Powder at 20°C
Nonacosanoic Acid
Safety and Handling

Safety and Handling

EINECSNumber : (2)-0608

TSCA : No