missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nonacosanoic Acid 95.0+%, TCI America™
Supplier: TCI America N06625G
Specifications
Nonacosanoic Acid | |
90°C | |
C29H58O2 | |
5 g | |
CCCCCCCCCCCCCCCCCCCCCCCCCCCCC(=O)O | |
438.781 | |
CHEBI:84867 | |
≥95.0% (GC) |
4250-38-8 | |
White | |
MFCD00057830 | |
IHEJEKZAKSNRLY-UHFFFAOYSA-N | |
nonacosanoic acid | |
20245 | |
438.78 | |
Crystalline Powder |
Chemical Identifiers
4250-38-8 | |
438.781 | |
IHEJEKZAKSNRLY-UHFFFAOYSA-N | |
CHEBI:84867 | |
CCCCCCCCCCCCCCCCCCCCCCCCCCCCC(=O)O |
C29H58O2 | |
MFCD00057830 | |
20245 | |
nonacosanoic acid |
Safety and Handling
EINECSNumber : (2)-0608
TSCA : No