missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nonacarbonyldiiron, 99%

Manufacturer:  Alfa Aesar™ 1306209

 View more versions of this product

Catalog No. AA1306209



Specifications

Specifications

Nonacarbonyldiiron
Air Sensitive
~100°C (decomposition)
15321-51-4
Fe2(CO)9
UN3179
CGJSNKHFIBINMR-UHFFFAOYSA-N
carbon monoxide;iron;iron(3+);methanone
71311424
Crystalline
2.85g/mL
Orange
10g
C9H3Fe2O9
MFCD00151465
iron nonacarbonyl, enneacarbonyldiiron, nonacarbonyldiiron, 1,3-diferrabicyclo 1.1.1 pentane-2,4,5-trione; hexakis carbon monoxide, diironnonacarbonyl, diironnonacarbonyl, purum fe
[CH-]=O.[CH-]=O.[CH-]=O.[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Fe].[Fe+3]
366.804
363.79
99%

Chemical Identifiers

15321-51-4
366.804
CGJSNKHFIBINMR-UHFFFAOYSA-N
71311424
[CH-]=O.[CH-]=O.[CH-]=O.[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[C-]#[O+].[Fe].[Fe+3]
C9H3Fe2O9
MFCD00151465
iron nonacarbonyl, enneacarbonyldiiron, nonacarbonyldiiron, 1,3-diferrabicyclo 1.1.1 pentane-2,4,5-trione; hexakis carbon monoxide, diironnonacarbonyl, diironnonacarbonyl, purum fe
carbon monoxide;iron;iron(3+);methanone
Safety and Handling

Safety and Handling

GHS H Statement
H228-H301-H330
Flammable solid.
Toxic if swallowed.
Fatal if inhaled.

GHS P Statement
P210-P280f-P304+P340-P310
Keep away from heat/sparks/open flames/hot surfaces.
No smoking.
Wear protective clothing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 4.1; Packing Group: II; Proper Shipping Name: FLAMMABLE SOLID, TOXIC, INORGANIC, N.O.S.

TSCA : TSCA

Documents