missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Greiner Bio-One Non-Vented Polystyrene Petri Dishes

Used for easy stacking

Supplier:  Greiner Bio-One 632102

 View more versions of this product

Catalog No. 07000325


Explore available promotions

Description

Description

  • For long incubation periods
  • Manufactured from crystal polystyrene for optimal analysis by microscopy
Specifications

Specifications

80 mL
Polystyrene
Non-sterile
480
No
94 mm
Clear
Round
Heavy Design
20
16 mm
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.