missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Greiner Bio-One Non-Vented Polystyrene Petri Dishes

Used for easy stacking

Supplier:  Greiner Bio-One 632161

 View more versions of this product

Catalog No. 07000324



Description

Description

  • For long incubation periods
  • Manufactured from crystal polystyrene for optimal analysis by microscopy
Specifications

Specifications

80 mL
Polystyrene
Sterile
480
No
94 mm
Clear
Round
Heavy Design
20
16 mm
SDS
Documents

Documents

Promotions

Promotions

For Research Use Only. Not for use in diagnostic procedures, unless otherwise indicated.