missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nomega-(4-Methoxy-2,3,6-trimethylphenylsulfonyl)-L-arginine monohydrate, 95%

Manufacturer:  Alfa Aesar™ H6369703

 View more versions of this product

Catalog No. AAH6369703



Specifications

Specifications

Nomega-(4-Methoxy-2,3,6-trimethylphenylsulfonyl)-L-arginine monohydrate
C16H26N4O5S
h-arg mtr-oh, h-arg mtr-oh.1/2h2o, s-2-amino-5-3-4-methoxy-2,3,6-trimethylphenyl sulfonyl guanidino pentanoic acid, l-arginyl-4-methoxy-2,3,6-trimethylbenzenesulfonyl, n'-4-methoxy-2,3,6-trimethylbenzenesulfonyl-l-arginine, nomega-4-methoxy-2,3,6-trimethylbenzenesulfonyl-d-arginine, nomega-4-methoxy-2,3,6-trimethylbenzenesulfonyl-l-arginine, amino acid series, arginine mtr-oh, n pound inverted question mark 4-methoxy-2,3,6-trimethylbenzenesulfonyl-l-arginine
CC1=CC(=C(C(=C1S(=O)(=O)NC(=NCCCC(C(=O)O)N)N)C)C)OC
386.467
404.48
95%
1g
80745-10-4
MFCD01862361
FUSAEZSSVDNYPO-LBPRGKRZSA-N
(2S)-2-amino-5-[[amino-[(4-methoxy-2,3,6-trimethylphenyl)sulfonylamino]methylidene]amino]pentanoic acid
7408178
Powder
White
Moisture Sensitive

Chemical Identifiers

80745-10-4
386.467
FUSAEZSSVDNYPO-LBPRGKRZSA-N
7408178
CC1=CC(=C(C(=C1S(=O)(=O)NC(=NCCCC(C(=O)O)N)N)C)C)OC
C16H26N4O5S
MFCD01862361
h-arg mtr-oh, h-arg mtr-oh.1/2h2o, s-2-amino-5-3-4-methoxy-2,3,6-trimethylphenyl sulfonyl guanidino pentanoic acid, l-arginyl-4-methoxy-2,3,6-trimethylbenzenesulfonyl, n'-4-methoxy-2,3,6-trimethylbenzenesulfonyl-l-arginine, nomega-4-methoxy-2,3,6-trimethylbenzenesulfonyl-d-arginine, nomega-4-methoxy-2,3,6-trimethylbenzenesulfonyl-l-arginine, amino acid series, arginine mtr-oh, n pound inverted question mark 4-methoxy-2,3,6-trimethylbenzenesulfonyl-l-arginine
(2S)-2-amino-5-[[amino-[(4-methoxy-2,3,6-trimethylphenyl)sulfonylamino]methylidene]amino]pentanoic acid
Safety and Handling

Safety and Handling

storageNote1 : 2° to 8°C

SDS Documents