Please login to your online account to display your discounted pricing

Alfa Aesar™ Nomega-(4-Methoxy-2,3,6-trimethylphenylsulfonyl)-L-arginine monohydrate, 95%

Chemical Identifiers

Chemical Name or Material Nomega-(4-Methoxy-2,3,6-trimethylphenylsulfonyl)-L-arginine monohydrate
CAS 80745-10-4
Formula Weight 404.48
MDL Number MFCD01862361
Synonym H-Arg(Mtr)-OH.H2O
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAH6369703

  SDS
alfa aesar™
H6369703
1g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAH6369706

  SDS
alfa aesar™
H6369706
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 404.48
MDL Number MFCD01862361
Assay 95%
Physical Form Powder
Synonym H-Arg(Mtr)-OH.H2O
Quantity 1g
Chemical Name or Material Nomega-(4-Methoxy-2,3,6-trimethylphenylsulfonyl)-L-arginine monohydrate
CAS 80745-10-4
Storage Note 1 2° to 8°C
Sensitivity Moisture Sensitive
Color White
Safety and Handling

2° to 8°C