Please login to your online account to display your discounted pricing

NNC 711, Tocris

Selective inhibitor of GAT-1

$239.63 - $976.15

Chemical Identifiers

Chemical Name or Material NNC 711
Assay Percent Range >99%
Molecular Formula C21H22N2O3·HCl
Formula Weight 386.88
Synonym 1,2,5,6-Tetrahydro-1-[2-[[(diphenylmethylene)amino]oxy]ethyl]-3-pyridinecarboxylic acid hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

177910

 
r & d systems
RDS177910
10mg Each for $239.63

177950

 
r & d systems
RDS177950
50mg Each for $976.15
Description & Specifications

Specifications

Formula Weight 386.88
Quantity 10mg
Chemical Name or Material NNC 711
Assay Percent Range >99%
Recommended Storage Desiccate at +4°C
Synonym 1,2,5,6-Tetrahydro-1-[2-[[(diphenylmethylene)amino]oxy]ethyl]-3-pyridinecarboxylic acid hydrochloride
Molecular Formula C21H22N2O3·HCl
CAS 145645-62-1
Safety and Handling

Desiccate at +4°C