Please login to your online account to display your discounted pricing

NNC 63-0532, Tocris

Potent non-peptide NOP agonist; brain penetrant

$270.88 - $1,140.36

Chemical Identifiers

Chemical Name or Material NNC 63-0532
Assay Percent Range >98%
Molecular Formula C27H29N3O3
Formula Weight 443.54
Synonym 8-(1-Naphthalenylmethyl)-4-oxo-1-phenyl-1,3,8-triazaspiro[4.5]decane-3-acetic acid, methyl ester
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

178010

 
r & d systems
RDS178010
10mg Each for $270.88

178050

 
r & d systems
RDS178050
50mg Each for $1,140.36
Description & Specifications

Specifications

Formula Weight 443.54
Quantity 10mg
Chemical Name or Material NNC 63-0532
Assay Percent Range >98%
Molecular Formula C27H29N3O3
CAS 250685-44-0
Synonym 8-(1-Naphthalenylmethyl)-4-oxo-1-phenyl-1,3,8-triazaspiro[4.5]decane-3-acetic acid, methyl ester
Recommended Storage Store at Room Temperature
Safety and Handling

Store at Room Temperature