Please login to your online account to display your discounted pricing

NNC 05-2090 hydrochloride, Tocris

GABA uptake inhibitor; moderately BGT-1 selective

$257.85 - $1,073.10

Chemical Identifiers

Chemical Name or Material NNC 05-2090 hydrochloride
Assay Percent Range >98%
Molecular Formula C27H30N2O2·HCl
Formula Weight 451.01
Synonym 1-[3-(9H-Carbazol-9-yl)propyl]-4-(2-methoxyphenyl)-4-piperidinol hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

274710

 
r & d systems
RDS274710
10mg Each for $257.85

274750

 
r & d systems
RDS274750
50mg Each for $1,073.10
Description & Specifications

Specifications

Formula Weight 451.01
Quantity 10mg
Chemical Name or Material NNC 05-2090 hydrochloride
Assay Percent Range >98%
Molecular Formula C27H30N2O2·HCl
CAS 184845-18-9
Synonym 1-[3-(9H-Carbazol-9-yl)propyl]-4-(2-methoxyphenyl)-4-piperidinol hydrochloride
Recommended Storage Desiccate at Room Temperature
Safety and Handling

Desiccate at Room Temperature