Please login to your online account to display your discounted pricing

NMR Reference Standards, 5mm OD, Wilmad Bel-Art™ SP Scienceware™

Packaged in ultra-high field 8 in. length precision tubes. NMR Reference Standards, 5mm OD, Wilmad Bel-Art™ SP Scienceware™ guarantee their performance in experiments over 600MHz, comes with certificate of analysis (CofA).

$378.34 - $670.71

Chemical Identifiers

Packaging Ultra-high field precision tubes
Optical Density 5mm
Sensitivity 1H Sensitivity
Certificate of Analysis Included
Description 8 in. length tubes, 600MHz
Solution Type NMR reference standard solution
View More Specs
Products
Catalog Number Mfr. No. Product Type Solution Contains Quantity Price Quantity    

16-800-189

 
bel art products
660000089
Model: WG-R-01-5 0.1% ethylbenzene in chloroform-d Each Each for $520.94

16-800-190

 
bel art products
660000090
Model: WG-R-02-5 3% CHCI3 / 0.2% TMS in acetone-d6 Each Each for $541.34

16-800-191

 
bel art products
660000091
Model: WG-R-03-5 1% CHCI3 in acetone-d6 Each Each for $378.34

16-800-193

 
bel art products
660000093
Model: WG-R-13-5 0.05% Trifluorotoluene in CDCI3 Each Each for $670.71
Description & Specifications

Specifications

Concentration or Composition (by Analyte or Components) 0.1% ethylbenzene in chloroform-d
Quantity Each
Description 8 in. length tubes, 600MHz
Optical Density 5mm
Product Type Model: WG-R-01-5
Solution Type NMR reference standard solution
Packaging Ultra-high field precision tubes
Sensitivity 1H Sensitivity
Solution Contains 0.1% ethylbenzene in chloroform-d
Certificate of Analysis Included