Please login to your online account to display your discounted pricing

NMDA, Tocris


Selective NMDA agonist

Quantity 50mg

Manufacturer: r & d systems  RDS011450

 View more versions of this product

Catalog No. 011450

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 147.13
Assay Percent Range >99%
Molecular Formula C5H9NO4
Synonym N-Methyl-D-aspartic acid
Chemical Name or Material NMDA
CAS 6384-92-5
Recommended Storage Store at Room Temperature

Specifications

Chemical Name or Material NMDA
Assay Percent Range >99%
Molecular Formula C5H9NO4
Formula Weight 147.13
Synonym N-Methyl-D-aspartic acid
Accessories

Store at Room Temperature