Invitrogen™ NLK Recombinant Human Protein DFS Item
NLK is a serine/threonine kinase.
Manufacturer:  Invitrogen™ LSPV4328
 View more versions of this product
Catalog No. PV4328
Description
Description
NLK is involved in signal transduction and cellular communication.
Specifications
Specifications
NP_057315.1
Frozen
84.5kDa
Store at -80°C. At first use, aliquot and store at -80°C to avoid multiple freeze-thaws.
Full Length
Kinase Assay
MAP Kinase Family
Yes
Protein Biology|Enzyme & Protein Activity Assays|Kinase Activity|Protein Assays and Analysis
50mM Tris-HCl pH 7.5, 150mM NaCl, 0.5mM EDTA, 0.02% Triton X-100, 2mM DTT, 50% Glycerol
1mg
RUO - research use only
NLK
Baculovirus
Human
GST Tag
Documents