Please login to your online account to display your discounted pricing

Alfa Aesar™ [Nle&superscript_begin;13&superscript_end;]-Motillin

For cellular and molecular biology applications

Specifications

Product Type [Nle13]-Motillin
Molecular Weight 2681.10
Storage Requirements -30° to -10°C
Format Solid
Regulatory Status RUO
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66044EXD

 
alfa aesar™
J66044EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66044MCR

 
alfa aesar™
J66044MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type [Nle13]-Motillin
Molecular Weight 2681.10
Storage Requirements -30° to -10°C
Format Solid
Regulatory Status RUO
Purification Method HPLC