Please login to your online account to display your discounted pricing

Alfa Aesar™ [Nle&superscript_begin;13&superscript_end;, Glu&superscript_begin;14&superscript_end;] Motilin, Human, Porcine

For cellular and molecular biology applications

Specifications

Product Type [Nle13, Glu14] Motilin, human, porcine
Cross Reactivity Human, Ovine, Rat
Molecular Weight 2682.10
Storage Requirements -30° to -10°C
Sequence Phe-Val-Pro-Ile-Phe-Thr-Tyr-Gly-Glu-Leu-Gln-Arg-Nle-Glu-Glu-Lys-Glu-Arg-Asn-Lys-Gly-Gln
View More Specs

 Disclaimers

For Research Use Only

Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAJ66270EXD

 
alfa aesar™
J66270EXD
0.5mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAJ66270MCR

 
alfa aesar™
J66270MCR
1mg CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Product Type [Nle13, Glu14] Motilin, human, porcine
Cross Reactivity Human, Ovine, Rat
Molecular Weight 2682.10
Storage Requirements -30° to -10°C
Sequence Phe-Val-Pro-Ile-Phe-Thr-Tyr-Gly-Glu-Leu-Gln-Arg-Nle-Glu-Glu-Lys-Glu-Arg-Asn-Lys-Gly-Gln
Format Solid
Regulatory Status RUO
Purification Method HPLC