missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

NKH 477, Tocris

Water-soluble adenylyl cyclase activator

$272.44 - $1,145.83

Chemical Identifiers

Assay Percent Range >98%
Molecular Formula C27H43NO8·HCl
Formula Weight 546.1
Synonym N,N-Dimethyl-(3R,4aR,5S,6aS,10S,10aR,10bS)-5-(acetyloxy)-3-ethenyldodecahydro-10,10b-dihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-1H-naphtho[2,1-b]pyran-6-yl ester β-alanine hydrochloride
Chemical Name or Material NKH 477
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
160350 R & D Systems
RDS160350
50mg Each for $1,145.83
160310 R & D Systems
RDS160310
10mg Each for $272.44
Specifications Description & Specifications

Chemical Identifiers

>98%
546.1
NKH 477
C27H43NO8·HCl
N,N-Dimethyl-(3R,4aR,5S,6aS,10S,10aR,10bS)-5-(acetyloxy)-3-ethenyldodecahydro-10,10b-dihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-1H-naphtho[2,1-b]pyran-6-yl ester β-alanine hydrochloride

Specifications

138605-00-2
C27H43NO8·HCl
N,N-Dimethyl-(3R,4aR,5S,6aS,10S,10aR,10bS)-5-(acetyloxy)-3-ethenyldodecahydro-10,10b-dihydroxy-3,4a,7,7,10a-pentamethyl-1-oxo-1H-naphtho[2,1-b]pyran-6-yl ester β-alanine hydrochloride
NKH 477
>98%
546.1
50mg

Safety and Handling

Recommended Storage : Desiccate at +4°C