Please login to your online account to display your discounted pricing

NKG2AB6 Mouse anti-Mouse, PerCP-eFluor 710, Clone: 16a11, eBioscience™

Mouse Monoclonal Antibody

$187.05 - $419.05

Specifications

Antigen NKG2AB6
Clone 16a11
Host Species Murine
Species Reactivity Mouse
Applications Flow Cytometry
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

501123861

 
life technologies
LS46589780
25μg Each for $187.05

501129072

 
life technologies
LS46589782
100μg Each for $419.05
Description & Specifications

Specifications

Antigen NKG2AB6
Clone 16a11
Host Species Murine
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PerCP-eFluor 710
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material. This tandem dye is sensitive to photo-induced oxidation. Protect this vial from light during storage
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The 16a11 monoclonal antibody reacts with the mouse NKG2AB6. NKG2 molecules belong to a C-type lectin-like family of cell surface receptors expressed by mouse NK and NKT cell lineages. NKG2 molecules form heterodimeric complexes with CD95 and are responsible for recognition of non-classical MHC class I antigen Qa-1. This antibody reacts with the C57Bl/6 mouse strain, but does not react with the BALB/c or 129 mouse strains.

When co-staining with NKG2A/C/E clone 20d5, it is important to stain for NKG2AB6 first then subsequently with NKG2A/C/E, as steric hindrance has been observed if co-stained concurrently.