missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

NKG2A/C/E Rat anti-Mouse, Biotin, Clone: 20d5, eBioscience™

Rat Monoclonal Antibody

$91.08 - $157.08

Specifications

Antigen NKG2A/C/E
Clone 20d5
Host Species Rat
Gene Alias NKG2A, NKG2C, NKG2E
Applications Flow Cytometry
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5012016 Life Technologies
LS13589681
50μg Each for $91.08
5012017 Life Technologies
LS13589682
100μg Each for $157.08
Specifications

Description

The 20d5 monoclonal antibody reacts with the mouse NKG2A, C, and E. NKG2 molecules belong to a C-type lectin-like family of cell surface receptors expressed by mouse NK and NKT cell lineages. NKG2 molecules form heterodimeric complexes with CD94 and are responsible for recognition of non-classical MHC class I antigen Qa-1.

When co-staining with NKG2AB6 clone 16a11, it is important to stain for NKG2A-B6 first then subsequently with NKG2A/C/E, as steric hindrance has been observed if co-stained concurrently.
Description & Specifications

Specifications

NKG2A/C/E
Rat
Flow Cytometry
Biotin
Store at 2-8°C. Do not freeze.
Monoclonal
0.5mg/mL
20d5
NKG2A, NKG2C, NKG2E
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer