missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

NK1.1, PE-eFluor 610, clone: PK136, eBioscience™

Mouse Monoclonal Antibody

Manufacturer:  Life Technologies LS61594180

 View more versions of this product

Catalog No. 501128604



Description

Description

Killer cell lectin-like receptor subfamily B, member 1, also known as KLRB1, NKR-P1A or CD161 (cluster of differentiation 161), is a human gene. Natural killer (NK) cells are lymphocytes that mediate cytotoxicity and secrete cytokines after immune stimulation. Several genes of the C-type lectin superfamily, including the rodent NKRP1 family of glycoproteins, are expressed by NK cells and may be involved in the regulation of NK cell function. The KLRB1 protein contains an extracellular domain with several motifs characteristic of C-type lectins, a transmembrane domain, and a cytoplasmic domain. The KLRB1 protein, NKR-P1A or CD161, is classified as a type II membrane protein because it has an external C terminus. NKR-P1A, the receptor encoded by the KLRB1 gene, recognizes Lectin Like Transcript-1 (LLT1) as a functional ligand.

Light sensitivity: This tandem dye is sensitive to photo-induced oxidation. Please protect this vial and stained samples from light. Fixation: Samples can be stored in IC Fixation Buffer (cat. 00-8222) (100 µL of cell sample + 100 µL of IC Fixation Buffer) or 1-step Fix/Lyse Solution (cat. 00-5333) for up to 3 days in the dark at 4°C with minimal impact on brightness and FRET efficiency/compensation. Some generalizations regarding fluorophore performance after fixation can be made, but clone specific performance should be determined empirically. Excitation: 488-561 nm; Emission: 607 nm; Laser: Blue Laser, Green Laser, Yellow-Green Laser. Filtration: 0.2 μm post-manufacturing filtered. NK1.1 (Killer cell lectin-like receptor subfamily B, member 1, KLRB1, NKR-P1A, CD161, cluster of differentiation 161), refers to Natural Killer (NK) cells, lymphocytes that mediate cytotoxicity and secrete cytokines after immune stimulation. Several genes of the C-type lectin superfamily, including the rodent NKRP1 family of glycoproteins, are expressed by NK cells and may be involved in the regulation of NK cell function. The KLRB1 protein contains an extracellular domain with several motifs characteristic of C-type lectins, a transmembrane domain, and a cytoplasmic domain. The KLRB1 protein, NKR-P1A or CD161, is classified as a type II membrane protein because it has an external C terminus. NKR-P1A, the receptor encoded by the KLRB1 gene, recognizes Lectin Like Transcript-1 (LLT1) as a functional ligand.
Specifications

Specifications

NK1.1
Monoclonal
0.2 mg/mL
PBS with 0.09% sodium azide; pH 7.2
P27814
Klrb1d, Ly55b, Ly55d, MGC163757, MGC163759, NKR-P1B, NKR-P1D, Nkrp1-b, Nkrp1d
Mouse
Affinity chromatography
RUO
Antibody
Monoclonal
Mouse
Flow Cytometry
PK136
PE-eFluor® 610
Klrb1c
Liquid
Klrb1c
IgG2a, kappa
25 μg
4° C, store in dark, DO NOT FREEZE!
Primary
17059
Documents

For Research Use Only.