Please login to your online account to display your discounted pricing

NK1.1 Mouse anti-Mouse, PE, Clone: PK136, eBioscience™

Mouse Monoclonal Antibody

$56.55 - $404.55

Specifications

Antigen NK1.1
Clone PK136
Host Species Murine
Gene Alias CD161, NKR-P1C, Ly-55
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5011000

 
life technologies
LS12594163
10μg Each for $56.55

5011001

 
life technologies
LS12594181
50μg Each for $129.05

5011003

 
life technologies
LS12594183
200μg Each for $404.55

501129610

 
life technologies
LS12594182
100μg Each for $259.55
Description & Specifications

Specifications

Antigen NK1.1
Clone PK136
Host Species Murine
Gene Alias CD161, NKR-P1C, Ly-55
Species Reactivity Mouse
Applications Flow Cytometry
Regulatory Status RUO
Conjugate PE
Format Conjugated
Storage Requirements Store at 2-8°C. Do not freeze. Light-sensitive material.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
Concentration 0.2mg/mL

The PK136 monoclonal antibody reacts with mouse NK1.1, an antigen expressed by natural killer cells and a subset of T cells in the NK1.1 mouse strains including C57BL and NZB. Several commonly used laboratory mouse strains such as BALB/c, SJL, AKR, CBA, C3H and A do not express the NK1.1 antigen. For detection of NK cells in these strains the monoclonal antibody DX5 (Cat. No. 14-5971) should be used. Simultaneous staining of C57BL/6 spleen cells with PK136 and DX5 reveals coexpression of both markers by a majority of cells as well as presence of small populations of DX5+PK136- and DX5-PK136+ cells.