missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

NK1.1 Mouse anti-Mouse, eFluor 450, Clone: PK136, eBioscience™

Mouse Monoclonal Antibody

$132.00 - $295.68

Specifications

Antigen NK1.1
Clone PK136
Host Species Murine
Gene Alias CD161, NKR-P1C, Ly-55
Species Reactivity Mouse
View More Specs

Disclaimers:

For Research Use Only.



Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5016387 Life Technologies
LS48594180
25μg Each for $132.00
5016388 Life Technologies
LS48594182
100μg Each for $295.68
Specifications

Description

The PK136 monoclonal antibody reacts with mouse NK1.1, an antigen expressed by natural killer cells and a subset of T cells in the NK1.1 mouse strains including C57BL and NZB. Several commonly used laboratory mouse strains such as BALB/c, SJL, AKR, CBA, C3H and A do not express the NK1.1 antigen. For detection of NK cells in these strains the monoclonal antibody DX5 (Cat. No. 14-5971) should be used. Simultaneous staining of C57BL/6 spleen cells with PK136 and DX5 reveals coexpression of both markers by a majority of cells as well as presence of small populations of DX5+PK136- and DX5-PK136+ cells.
Description & Specifications

Specifications

NK1.1
Murine
Mouse
RUO
Conjugated
Primary
aqueous buffer, 0.09% sodium azide, may contain carrier protein/stabilizer
PK136
CD161, NKR-P1C, Ly-55
Flow Cytometry
eFluor 450
Store at 2-8°C. Do not freeze. Light-sensitive material.
Monoclonal
0.2mg/mL