Please login to your online account to display your discounted pricing

NK1.1 Mouse anti-Mouse, Clone: PK136, Functional Grade, eBioscience™

Mouse Monoclonal Antibody, Functional Grade

$114.55 - $433.55

Specifications

Antigen NK1.1
Clone PK136
Host Species Mouse
Gene Alias CD161, NKR-P1C, Ly-55
Species Reactivity Mouse
View More Specs

 Disclaimers

For Research Use Only.

Products
Catalog Number Mfr. No. Quantity Price Quantity    

5014641

 
life technologies
LS16594181
50μg Each for $114.55

5014642

 
life technologies
LS16594182
100μg Each for $216.05

5014643

 
life technologies
LS16594185
500μg Each for $433.55
Description & Specifications

Specifications

Antigen NK1.1
Clone PK136
Host Species Mouse
Gene Alias CD161, NKR-P1C, Ly-55
Species Reactivity Mouse
Applications Cell Depletion
Applications Flow Cytometry
Applications Functional Assay
Regulatory Status RUO
Conjugate Unlabeled
Format Purified
Storage Requirements Store at 2-8°C.
Primary or Secondary Primary
Monoclonal or Polyclonal Monoclonal
Formulation aqueous buffer, no sodium azide
Concentration 1mg/mL

The PK136 monoclonal antibody reacts with mouse NK1.1, an antigen expressed by natural killer cells and a subset of T cells in the NK1.1 mouse strains including C57BL and NZB. Several commonly used laboratory mouse strains such as BALB/c, SJL, AKR, CBA, C3H and A do not express the NK1.1 antigen. For detection of NK cells in these strains the monoclonal antibody DX5 (Cat. No. 14-5971) should be used. Simultaneous staining of C57BL/6 spleen cells with PK136 and DX5 reveals coexpression of both markers by a majority of cells as well as presence of small populations of DX5+PK136- and DX5-PK136+ cells.