Please login to your online account to display your discounted pricing

Alfa Aesar™ Nitrosylated Endothelial Nitric Oxide Synthase (eNOS)(194-204), Certified Mass Spec Grade


For cellular and molecular biology applications

Manufacturer: alfa aesar™  J67614ODL

 View more versions of this product

 Disclaimers

For Research Use Only

Catalog No. AAJ67614ODL

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Specifications

Regulatory Status RUO
Molecular Weight 1297.40
Quantity 3 vials, 500 pmol/vial
Sequence VFDARD-C(NO)-RSAQ
Purification Method HPLC
Storage Requirements -30° to -10°C
Format Lyophilized powder
Product Type Nitrosylated Endothelial Nitric Oxide Synthase (eNOS) (194-204), Certified Mass Spec Grade