missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nitrosonium hexafluorophosphate, 96%

Manufacturer:  Alfa Aesar™ 1314906

 View more versions of this product

Catalog No. AA1314906



Specifications

Specifications

Nitrosonium hexafluorophosphate
Moisture Sensitive
16921-91-8
NOPF6
UN3260
SAKPNYRUBHJGAR-UHFFFAOYSA-N
azanylidyneoxidanium;hexafluorophosphate
10910083
Crystalline
Odorless
5g
F6NOP
MFCD00040324
nitrosonium hexafluorophosphate, nitrilooxonium hexafluorophosphate, nopf6, nitrosyl hexafluorophosphate, nitrosonium hexafluorophosphat, nitrosylhexafluorophosphate, azanylidyneoxidanium hexafluorophosphate, hexafluoro-$l^ 5-phosphanuide; nitrosonium
N#[O+].F[P-](F)(F)(F)(F)F
174.97
174.96
96%

Chemical Identifiers

16921-91-8
174.97
SAKPNYRUBHJGAR-UHFFFAOYSA-N
10910083
N#[O+].F[P-](F)(F)(F)(F)F
F6NOP
MFCD00040324
nitrosonium hexafluorophosphate, nitrilooxonium hexafluorophosphate, nopf6, nitrosyl hexafluorophosphate, nitrosonium hexafluorophosphat, nitrosylhexafluorophosphate, azanylidyneoxidanium hexafluorophosphate, hexafluoro-$l^ 5-phosphanuide; nitrosonium
azanylidyneoxidanium;hexafluorophosphate
Safety and Handling

Safety and Handling

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing.
Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF SWALLOWED: Rinse mouth.
Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, INORGANIC, N.O.S.

TSCA : TSCA

SDS Documents