missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nitronium tetrafluoroborate, 97%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals 174380050
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nitronium tetrafluoroborate | |
Colorless through White | |
Adhering Crystals | |
96% min | |
NO2BF4 | |
01,742; 04,358; 05,476; 08,361; 09,324; 01,747; 04,360; 07,253; 08,364; 14,215 | |
Solubility in water: may decompose | |
[O-][NH+]=O.F[B-](F)(F)F | |
132.81 | |
132.81 | |
Plastic bottle |
200.0°C | |
13826-86-3 | |
5 g | |
BF4NO2 | |
MFCD00011432 | |
nitronium tetrafluoroborate, nitronium ion tetrafluoroborate, nitronium tetrafluoroboron, nitroniumtetrafluoroborate, nitronium.tetrafluoroborate, nitronium tetrafluoroborate, 0.3-0.5m solution in sulfolane | |
XYRQURYIJKJVSF-UHFFFAOYSA-N | |
nitronium;tetrafluoroborate | |
11073463 | |
97% |
Chemical Identifiers
13826-86-3 | |
132.81 | |
XYRQURYIJKJVSF-UHFFFAOYSA-N | |
11073463 | |
[O-][NH+]=O.F[B-](F)(F)F |
BF4NO2 | |
MFCD00011432 | |
nitronium tetrafluoroborate, nitronium ion tetrafluoroborate, nitronium tetrafluoroboron, nitroniumtetrafluoroborate, nitronium.tetrafluoroborate, nitronium tetrafluoroborate, 0.3-0.5m solution in sulfolane | |
nitronium;tetrafluoroborate |
Safety and Handling
GHS Signal Word: Danger
EINECSNumber : 237-533-5
RUO – Research Use Only