Please login to your online account to display your discounted pricing

Alfa Aesar™ Nitronium hexafluoroantimonate(V), 95%

Chemical Identifiers

Chemical Name or Material Nitronium hexafluoroantimonate(V)
CAS 17856-92-7
Molecular Formula NO2SbF6
Formula Weight 281.75g/mol
MDL Number MFCD00042551
UN Number UN1549
Synonym Nitryl hexafluoroantimonate
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAB2092406

 
alfa aesar™
B20924-06
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2092414

 
alfa aesar™
B20924-14
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAB2092422

 
alfa aesar™
B20924-22
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Quantity 5g
Chemical Name or Material Nitronium hexafluoroantimonate(V)
CAS 17856-92-7
Assay 95%
Molecular Formula NO2SbF6
Formula Weight 281.75g/mol
MDL Number MFCD00042551
UN Number UN1549
Synonym Nitryl hexafluoroantimonate
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.
Health Hazard 3 GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.
Recommended Storage −30° to −10°C
DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ANTIMONY COMPOUNDS, INORGANIC, SOLID, N.O.S.
Safety and Handling

Warning

GHS P Statement
P261-P264-P304+P340-P301+P312-P312-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wash thoroughly after handling.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Call a POISON CENTER or doctor/physician if you feel unwell.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H302-H332
Harmful if swallowed.
Harmful if inhaled.

−30° to −10°C