Please login to your online account to display your discounted pricing

Alfa Aesar™ Nitromethane-d3, 99%(Isotopic)

Chemical Identifiers

Chemical Name or Material Nitromethane-d3
CAS 13031-32-8
Name Note Isotopic
Formula Weight 64.06
MDL Number MFCD00044214
UN Number UN1261
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4233806

  SDS
alfa aesar™
4233806
5g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AA4233814

  SDS
alfa aesar™
4233814
25g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Formula Weight 64.06
Chemical Name or Material Nitromethane-d3
TSCA TSCA
Density 1.183g/mL
Name Note Isotopic
Boiling Point 100°C
Physical Form Liquid
Assay 99%
Quantity 5g
Color Colorless
Refractive Index 1.3795
MDL Number MFCD00044214
CAS 13031-32-8
UN Number UN1261
Health Hazard 1 Warning
Health Hazard 2 GHS H Statement
H226-H302
Flammable liquid and vapor.
Harmful if swallowed.
Odor Characteristic
Health Hazard 3 GHS P Statement
P260-P370+P380+P375
Do not breathe dust/fume/gas/mist/vapors/spray.
In case of fire: Evacuate area.
Fight fire remotely due to the risk of explosion.
DOT Information Transport Hazard Class: 3; Packing Group: II; Proper Shipping Name: NITROMETHANE
Safety and Handling

Warning

GHS P Statement
P260-P370+P380+P375
Do not breathe dust/fume/gas/mist/vapors/spray.
In case of fire: Evacuate area.
Fight fire remotely due to the risk of explosion.

GHS H Statement
H226-H302
Flammable liquid and vapor.
Harmful if swallowed.