Please login to your online account to display your discounted pricing

Nitromethane, ≥98%, Alfa Aesar™

Chemical Identifiers

Chemical Name or Material Nitromethane
CAS 75-52-5
Physical Form Liquid
Assay Percent Range ≥98%
Molecular Formula CH3NO2
Formula Weight 61.04
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AAA1180636

  SDS
alfa aesar
A1180636
500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA118060E

  SDS
alfa aesar
A118060E
2500g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

AAA1180622

  SDS
alfa aesar
A1180622
100g CALL N/A N/A
 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Description & Specifications

Specifications

Chemical Name or Material Nitromethane
Density 1.130
Quantity 500g
CAS 75-52-5
Physical Form Liquid
Assay Percent Range ≥98%
Molecular Formula CH3NO2
Formula Weight 61.04

Applications
Nitromethane is used as a stabilizer of halogenated organic solvents, rocket and racing fuel and a chemical intermediate. It is also used as a solvent for cyanoacrylate adhesives, polymers and waxes. It serves as a Michael donor, adding to alfa,beta-unsaturated carbonyl compounds through 1,4-addition in the Michael reaction. It acts as a solvent used for extractions, reaction medium and as a cleaning solvent. Further, it is used in the manufacture of pharmaceuticals, explosives, fibers and coatings.

Solubility
Miscible with ethanol, ethyl ether, acetone, carbon tetrachloride and alkali. Slightly miscible with water.

Notes
Incompatible with amines, strong acids, strong bases, strong oxidizing agents, strong reducing agents and copper