missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Nitrogen Standard 5460, Ricca Chemical

Chemical Identifiers

CAS 7732-18-5,7632-00-0
CAS Min % 98.0,0.13
CAS Max % 100.0,0.13
Chemical Name or Material Nitrogen Standard, 1 mL = 0.25 mg N as NO2
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
5460-16 Ricca Chemical Company
RI546016
16 oz. N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
5460-4 Ricca Chemical Company
RI54604
4 oz. N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

7732-18-5,7632-00-0
98.0,0.13
100.0,0.13
Nitrogen Standard, 1 mL = 0.25 mg N as NO2

Specifications

16 oz.
7632-00-0,7732-18-5
Nitrogen Standard, 1 mL = 0.25 mg N as NO2
0.13,98.0
0.13,100.0