Learn More
Nitrocyclopentane, 99%, Thermo Scientific Chemicals
Supplier: Thermo Scientific Chemicals L0802906
Description
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Nitrocyclopentane | |
1.082 | |
67°C (152°F) | |
1.454 | |
5 g | |
cyclopentane, nitro, nitrocyclopentase, ccris 5057, nitro-cyclopentane, acmc-20ak2p, nitrocyclopentane, cjszwogckkdsjg-uhfffaoysa | |
[O-][N+](=O)C1CCCC1 | |
115.13 | |
115.13 |
2562-38-1 | |
179°C to 180°C | |
C5H9NO2 | |
MFCD00001362 | |
907079 | |
CJSZWOGCKKDSJG-UHFFFAOYSA-N | |
nitrocyclopentane | |
75714 | |
99% |
Chemical Identifiers
2562-38-1 | |
115.13 | |
CJSZWOGCKKDSJG-UHFFFAOYSA-N | |
75714 | |
[O-][N+](=O)C1CCCC1 |
C5H9NO2 | |
MFCD00001362 | |
cyclopentane, nitro, nitrocyclopentase, ccris 5057, nitro-cyclopentane, acmc-20ak2p, nitrocyclopentane, cjszwogckkdsjg-uhfffaoysa | |
nitrocyclopentane |
Safety and Handling
GHS H Statement
H301-H227-H332
Toxic if swallowed.
Combustible liquid.
Harmful if inhaled.
P210-P235-P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H302+H312+H332-H315-H319-H335
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 219-884-6
RTECSNumber : GY4657500
TSCA : No
Recommended Storage : Ambient temperatures
RUO – Research Use Only