Please login to your online account to display your discounted pricing

Nitrocyclopentane, +97%, ACROS Organics™


Quantity 1mL
Packaging Glass bottle

Manufacturer: thermo fisher scientific  AC381750010

 View more versions of this product

Catalog No. AC381750010

 Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.


Description & Specifications

Chemical Identifiers

Assay ≥97.0%
Physical Form Liquid
Formula Weight 115.13
Quantity 1mL
Color Clear pale yellow
Density 1.0860g/mL
CAS Max % 100.0
CAS Min % 97.0
Linear Formula C5H9NO2
Health Hazard 3 GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
Health Hazard 2 GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Molecular Formula C5H9NO2
Assay Percent Range 97+%
Infrared Spectrum Authentic
Packaging Glass bottle
Refractive Index 1.4530 to 1.4550 (20°C, 589nm)
Health Hazard 1 GHS Signal Word: Warning
CAS 2562-38-1
MDL Number MFCD00001362
Chemical Name or Material Nitrocyclopentane

Specifications

Chemical Name or Material Nitrocyclopentane
CAS 2562-38-1
CAS Min % 97.0
CAS Max % 100.0
Assay ≥97.0%
Assay Percent Range 97+%
Molecular Formula C5H9NO2
Linear Formula C5H9NO2
Formula Weight 115.13
MDL Number MFCD00001362
Accessories

GHS Signal Word: Warning

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if inhaled.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.

SDS & Safety Documents