Please login to your online account to display your discounted pricing

Nitrocaramiphen hydrochloride, Tocris

Muscarinic antagonist, M1 > M2

$163.07 - $678.85

Chemical Identifiers

Chemical Name or Material Nitrocaramiphen hydrochloride
Assay Percent Range >98%
Molecular Formula C18H26N2O4·HCl
Formula Weight 370.88
Synonym 2-Diethylaminoethyl 1-(4-nitrophenyl)cyclopentanecarboxylate hydrochloride
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

046910

 
r & d systems
RDS046910
10mg Each for $163.07

046950

 
r & d systems
RDS046950
50mg Each for $678.85
Description & Specifications

Specifications

Quantity 10mg
Formula Weight 370.88
Molecular Formula C18H26N2O4·HCl
CAS 98636-73-8
Chemical Name or Material Nitrocaramiphen hydrochloride
Recommended Storage Store at Room Temperature
Assay Percent Range >98%
Synonym 2-Diethylaminoethyl 1-(4-nitrophenyl)cyclopentanecarboxylate hydrochloride
Safety and Handling

Store at Room Temperature