Please login to your online account to display your discounted pricing

Nitrocaramiphen hydrochloride, Tocris


Muscarinic antagonist, M1 > M2

Quantity 50mg

Manufacturer: r & d systems  RDS046950

 View more versions of this product

Catalog No. 046950

  • / Each
 

Description & Specifications

Chemical Identifiers

Quantity 50mg
Formula Weight 370.88
Recommended Storage Store at Room Temperature
Synonym 2-Diethylaminoethyl 1-(4-nitrophenyl)cyclopentanecarboxylate hydrochloride
Chemical Name or Material Nitrocaramiphen hydrochloride
Assay Percent Range >98%
CAS 98636-73-8
Molecular Formula C18H26N2O4·HCl

Specifications

Chemical Name or Material Nitrocaramiphen hydrochloride
Assay Percent Range >98%
Molecular Formula C18H26N2O4·HCl
Formula Weight 370.88
Synonym 2-Diethylaminoethyl 1-(4-nitrophenyl)cyclopentanecarboxylate hydrochloride
Accessories

Store at Room Temperature