Please login to your online account to display your discounted pricing

Alfa Aesar™ Nitrobenzene-d5, 99% (Isotopic)

$64.34 - $64.34

Chemical Identifiers

Chemical Name or Material Nitrobenzene-d5
CAS 4165-60-0
Name Note Isotopic
Formula Weight 128.14
MDL Number MFCD00044415
UN Number UN1662
View More Specs
Products
Catalog Number Mfr. No. Quantity Price Quantity    

AA4233706

 
alfa aesar™
4233706
5g Each for $64.34
Description & Specifications

Specifications

Formula Weight 128.14
Chemical Name or Material Nitrobenzene-d5
Density 1.25g/mL
Name Note Isotopic
Boiling Point 211°C
Melting Point 6°C
Physical Form Liquid
Assay 99%
Quantity 5g
Flash Point 88°C (190°F)
MDL Number MFCD00044415
CAS 4165-60-0
UN Number UN1662
Health Hazard 1 Danger
Health Hazard 2 GHS H Statement
H300-H310-H330-H372-H351-H361-H227
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
Combustible liquid.
Health Hazard 3 GHS P Statement
P210-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.
DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITROBENZENE
Safety and Handling

Danger

GHS P Statement
P210-P301+P310-P304+P340-P320-P330-P361-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Specific treatment is urgent (see label).
Rinse mouth.
Remove/Take off immediately all contaminated clothing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

GHS H Statement
H300-H310-H330-H372-H351-H361-H227
Fatal if swallowed.
Fatal in contact with skin.
Fatal if inhaled.
Causes damage to organs through prolonged or repeated exposure.
Suspected of causing cancer.
Suspected of damaging fertility or the unborn child.
Combustible liquid.