missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Please login to your online account to display your discounted pricing
Nitrobenzene (Certified ACS), Fisher Chemical™
Supplier: Fisher Chemical N91I4
Specifications
Nitrobenzene | |
5°C | |
1.205g/cm³ | |
Amber Glass/Poison Pack, EcoSafPak™ | |
MFCD00007043 | |
0.005% max. | |
LQNUZADURLCDLV-UHFFFAOYSA-N | |
nitrobenzene | |
4.25g/mol | |
CHEBI:27798 | |
0.2mbar at 20°C | |
Certified ACS |
98-95-3 | |
Yellow | |
210°C | |
C6H5NO2 | |
4 L | |
nitrobenzol, benzene, nitro, oil of mirbane, mirbane oil, essence of mirbane, nitro-benzene, nitrobenzeen, oil of myrbane, nitrobenzen, mononitrobenzene | |
C1=CC=C(C=C1)[N+](=O)[O-] | |
123.111 | |
7416 | |
123.11 | |
≥99.0% | |
Liquid |
Chemical Identifiers
98-95-3 | |
123.111 | |
LQNUZADURLCDLV-UHFFFAOYSA-N | |
7416 | |
nitrobenzene |
C6H5NO2 | |
MFCD00007043 | |
nitrobenzol, benzene, nitro, oil of mirbane, mirbane oil, essence of mirbane, nitro-benzene, nitrobenzeen, oil of myrbane, nitrobenzen, mononitrobenzene | |
CHEBI:27798 | |
C1=CC=C(C=C1)[N+](=O)[O-] |
Safety and Handling
DOTInformation : DOT Class 6.1, : Poison