Please login to your online account to display your discounted pricing

Nitrobenzene (Certified ACS), Fisher Chemical

$672.08 - $3,865.25

Chemical Identifiers

Chemical Name or Material Nitrobenzene
CAS 98-95-3
Purity Grade Certified ACS Grade
Assay ≥99.0%
Molecular Formula C6H5NO2
Formula Weight 123.11
MDL Number MFCD00007043
Synonym Mirbane Oil
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

N91I-4

  SDS
fisher scientific company
N91I4
Amber Glass/Poison Pack, EcoSafPak™ 4L Each for $3,865.25

N91I-500

  SDS
fisher scientific company
N91I500
Glass Bottle 500mL Each for $672.08
Description & Specifications

Specifications

Assay ≥99.0%
Melting Point 5°C
Purity Grade Certified ACS Grade
Formula Weight 123.11
Boiling Point 210°C
Quantity 4L
DOT Information DOT Class 6.1, : Poison
Physical Form Liquid
Density 1.205g/cm³
Vapor Pressure 0.2mbar at 20°C
Color Yellow
Vapor Density 4.25g/mol
Identification Pass Test
Packaging Amber Glass/Poison Pack, EcoSafPak™
Chloride 5ppm max.
ChemAlert Storage Symbol Blue
MDL Number MFCD00007043
Synonym Mirbane Oil
Residue after Evaporation 0.005% max.
Water_soluble Titratable Acid 0.0005mEq/g max.
CAS 98-95-3
Chemical Name or Material Nitrobenzene
Molecular Formula C6H5NO2

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

Safety and Handling