Please login to your online account to display your discounted pricing

Nitrobenzene, BAKER ANALYZED™ A.C.S. Reagent, J.T.Baker™

High quality chemicals for laboratory and specialized industrial use

$212.49 - $821.45

Chemical Identifiers

Chemical Name or Material Nitrobenzene
CAS 98-95-3
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula C6H5NO2
Formula Weight 123.11
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

02003524

 
avantor performance
932503
Each for $821.45

02003525

 
avantor performance
932501
Narrow Mouth Amber Glass Bottle 500mL Each for $212.49
Description & Specifications

Specifications

Chemical Name or Material Nitrobenzene
Flash Point 88.00°C
Specific Gravity 1L = 1.20kg
Quantity 4L
CAS 98-95-3
Purity Grade Baker Analyzed™ ACS Reagent
Molecular Formula C6H5NO2
Formula Weight 123.11