missing translation for 'onlineSavingsMsg'
Learn More
Please login to your online account to display your discounted pricing

Alfa Aesar™ Nitrite, Ion chromatography standard solution, Specpure™, NO2- 1000 μg/mL

Chemical Identifiers

MDL Number MFCD00144923
Chemical Name or Material Nitrite Ion chromatography standard solution, NO2- 1000μg/mL
Grade Specpure™
View More Specs


Products ${productFamilyLength}
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA40020AE SDS Alfa Aesar™
40020AE
100mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
AA40020AP SDS Alfa Aesar™
40020AP
500mL N/A N/A  
Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.
Specifications Description & Specifications

Chemical Identifiers

MFCD00144923
Nitrite Ion chromatography standard solution, NO2- 1000μg/mL
Specpure™

Specifications

Liquid
100mL
Specpure™
MFCD00144923
Nitrite Ion chromatography standard solution, NO2- 1000μg/mL

Safety and Handling

TSCA : TSCA