Please login to your online account to display your discounted pricing

Nitrite (as Nitrogen) Standard, 1000ppm, Preserved with Chloroform (1mL = 1mg N, 1mL = 3.28mg NO2-), Certified, LabChem™

$53.31 - $120.60

Chemical Identifiers

Chemical Name or Material Nitrite (as Nitrogen) Standard
CAS 7732-18-5
CAS Max % 99.19
CAS 7758-09-0
CAS Max % 0.61
CAS 67-66-3
CAS Max % 0.198
CAS 64-17-5
CAS Max % 0.002
Name Note 1000ppm, Preserved with Chloroform (1mL = 1mg N, 1mL = 3.28mg NO2-)
View More Specs
Products
Catalog Number Mfr. No. Packaging Quantity Price Quantity    

LC179301

 
lab chem
LC179301
Poly Bottle 500mL Each for $120.60

LC179307

 
lab chem
LC179307
Poly Bottle 125mL Each for $53.31
Description & Specifications

Specifications

Purity Grade Certified Grade
Formula Weight 85.1
Chemical Name or Material Nitrite (as Nitrogen) Standard
Specific Gravity 1g/mL
Color Clear, colorless
Quantity 500mL
Decomposition Information Nitrogen oxides; Hydrogen chloride; Carbon monoxide; Carbon dioxide
CAS Max % 0.198
CAS 64-17-5
CAS Max % 0.002
Name Note 1000ppm, Preserved with Chloroform (1mL = 1mg N, 1mL = 3.28mg NO2-)
Physical Form Liquid
Assay Percent Notes as N
Concentration 1000ppm ±10ppm (1mL = 1mg N, 1mL = 3.28mg NO2- )
Linear Formula KNO2
CAS 7732-18-5
Health Hazard 1 Warning
Identification Passes Test
CAS Max % 99.19
Health Hazard 2 GHS H Statement
Suspected of causing cancer.
Harmful to aquatic life.
CAS 7758-09-0
Health Hazard 3 GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Avoid release to the environment.
Wear protective gloves, eye protection.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.
Packaging Poly Bottle
Solubility Soluble in water
Traceability to NIST Traceable to NIST
CAS Max % 0.61
Recommended Storage Refrigerate
Density 1g/mL
CAS 67-66-3
Safety and Handling

Warning

GHS P Statement
Obtain special instructions before use.
Do not handle until all safety precautions have been read and understood.
Avoid release to the environment.
Wear protective gloves, eye protection.
If exposed or concerned: Get medical advice/attention.
Store locked up.
Dispose of contents/container to comply with local, state and federal regulations.

GHS H Statement
Suspected of causing cancer.
Harmful to aquatic life.

Refrigerate